Water for molecular biology, sterile, DEPC treated

Supplier: TEKNOVA CA

W0240 W0235
CA100218-178EA 436.25 CAD
CA100218-178 CA100219-828 CA100220-284
Water for molecular biology, sterile, DEPC treated
Water

DEPC-treated water is ideal for molecular biology applications involving RNA. This water is incubated with 0.1% diethyl pyrocarbonate (DEPC) to inactivate RNases and autoclaved twice to remove the DEPC and sterilize the final solution.


DEPC-treated water is not recommended for PCR or in vivo experiments.

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

DNase tested
RNase tested
Protease tested


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR