Water, Molecular Biology Grade, PCR certified DNase/RNase tested, sterile

Supplier: TEKNOVA CA

W3350 W3330 W3335 W3340 W3345
CA100216-650EA 276.07 CAD
CA100216-650 CA100219-594 CA100219-912 CA100219-942 CA100220-288
Water, Molecular Biology Grade, PCR certified DNase/RNase tested, sterile
Water

Molecular Biology Grade Water is PCR certified negative for DNAse and RNAse (sensitivity to femtomoles) and is negative for non-specific Endonuclease, Exonuclease and Protease activity.


  • Sterile

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

DNase tested
RNase tested
Protease tested
PCR tested


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR