Buffer, TBS Solution 10X Concentrate (TRIS Buffered Saline), G-Biosciences
Supplier: G-Biosciences
R029
R030
CA82021-504EA
261.1
CAD
CA82021-504
CA82021-506
Buffer, TBS Solution 10X Concentrate (TRIS Buffered Saline), G-Biosciences
Buffers
Electrophoresis Buffers
Washing Buffers
The TBS solution is primarily used as a wash and blocking buffer diluent for Western blot, ELISA and immunoassays.
10X TBS: Tris buffered saline [10X]: 100mM Tris HCl, 1.5M NaCl, pH 7.5