Buffer, PBS solution 10X concentrate (phosphate buffered saline), G-Biosciences
Supplier: G-Biosciences
R027
R028
CA82021-500EA
357.7
CAD
CA82021-500
CA82021-502
Buffer, PBS solution 10X concentrate (phosphate buffered saline), G-Biosciences
Buffers
Electrophoresis Buffers
Washing Buffers
10X PBS (Phosphate Buffered Saline) is a 10 fold concentrated, sterile, pH adjusted blend of phosphate buffer and saline solution, which when diluted 1 to 10 in ultrapure water produces a phosphate buffered saline of 8mM NaH2PO4, 150mM NaCl, 3mM KCl, and 2mM KH2PO4 at pH 7.4.
The 10X solution is a sterile solution.