D-(-)-Sorbitol solution 1 M
Supplier: G-Biosciences
R038
CA82023-110EA
161.56
CAD
CA82023-110
D-(-)-Sorbitol solution 1 M
D(-)-Sorbitol
Quality DNase and RNase free reagent solution for yeast research.
Formula:
HOH₂C(CH(OH))₄CH₂OH Storage Temperature: Refrigerator |
MDL Number:
MFCD00004708 CAS Number: 50-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...