Eppendorf® 5910 Ri Refrigerated Centrifuges
Supplier: Eppendorf
5943000131 5943000106 5943000114 5943000246 5943000343 5943000548
CA76478-968EA 23067.67 CAD
CA76478-968 CA76478-970 CA76478-972 CA76478-974 CA76478-976 CA76478-978
Eppendorf® 5910 Ri Refrigerated Centrifuges
Centrifuges Benchtop Centrifuges
The refrigerated Eppendorf Centrifuge 5910 Ri stands out with an intuitive touch screen interface and a range of innovative solutions for usability, documentation and user management. Its wide variety of fixed-angle and swing-bucket rotors accommodate a comprehensive spectrum of applications in a compact footprint.
  • Large seven inches VisioNize® touch interface for super-fast parameter settings via unique favorites function
  • User management with multiple authorization levels and documentation features supporting GLP/GxP requirements
  • Aerosol-tight Eppendorf QuickLock® caps and lids for fast and ergonomic lid locking
  • Quiet operation to improve work environment
  • Centrifuge lid with soft-touch lid closure
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 to 250 ml
Swing-bucket rotors and adapters for tubes and bottles from 0.2 to 1000 ml as well as all types of MTP, PCR or deepwell plates.
The refrigerated benchtop Centrifuge 5910 Ri stands out for its ease of use and versatility: The large seven inches VisioNize® touch interface with an intuitive user experience provides fast and error-free operation, ensuring efficient and reproducible runs. Additionally, the documentation function and user management support your quality control with multiple authorization levels, password protection, and documentation of all runs and user activities.
Together with a comprehensive portfolio of fixed-angle and swing-bucket rotors, this centrifuge accommodates a particularly broad spectrum of applications, making it the perfect choice for multiuser laboratories with diverse requirements.
A truly intuitive touch interface experience. With the unique favorites function, the setting of parameters is now faster than ever: Frequently-used settings for time, temperature, and speed are directly accessible on the home screen, allowing users to create individual runs with just three clicks. With the program function, up to 99 programs each consisting of all run parameters (incl. ramps and radius correction settings) with clearly assigned names and a rotor compatibility check can be saved to avoid operation errors.
Outstanding centrifuge rotor versatility for a broad range of applications: Cell harvests in bottles up to 1000 ml, large-scale DNA and RNA isolations and Ficoll® gradients for the purification of lymphocytes and monocytes are merely a few examples of the many possible areas of application for Centrifuge 5910 Ri. Its extensive versatility supports the needs of your lab today and well into the future.
Certifications: ACT Label Certificate, Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).