Manual/Press, Model 3622, Spex SamplePrep
Supplier: SPEX SamplePrep
3622
76182-384EA 21077.83 CAD
76182-384
Manual/Press, Model 3622, Spex SamplePrep
Presses
Full-size 25-ton (22.7 metric ton) Carver® hydraulic laboratory pellet press that accepts 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets.
  • Accepts all SPEX SamplePrep evacuable pellet die sets
  • Can be operated anywhere in the world, even under the most primitive conditions
  • Economical alternative to motorized presses for labs with a small sample load
  • Hardened steel buffer plate is engraved with concentric circles and enables easy centering of the die
  • Manual, hydraulic laboratory press with buffer plate
  • Rugged, reliable and noted for its extraordinary durability
  • Upper platen is equipped with a safety shield for operator protection
Manually controlled. Ideal for pressing sample pellets for XRF, IR, and other analytical techniques. Typical samples include: cement, rocks, minerals, soils, ceramics, pharmaceuticals
Certifications: CE Approved.