VWR® DuraChill® Recirculating Chillers, -10 to 70 °C, 120 V
Supplier: VWR International
CA02A1P1-41AV1N CA02A1T1-41AV1N CA03A1P1-41AV1N CA03A1T1-41AV1N CA05A1P1-41AV1N CA05A1T1-41AV1N CA10A3P2-41AV1N CA10A3T1-41AV1N
76472-556EA 11228.76 CAD
76472-556 76472-558 76472-564 76472-566 76472-572 76472-574 76472-580 76472-582
VWR® DuraChill® Recirculating Chillers, -10 to 70 °C, 120 V
Circulating Thermostats Closed Circulator Systems Cooling Circulators
DuraChill® Chillers provide stable and reliable cooling for various common heat removal applications, including laser etching, AA furnaces, ICP, rotary evaporators, vacuum systems, reaction vessels, plasma etching, and condenser cooling.
  • Large, full-color touchscreen display with intuitive interface and support for five different languages
  • Compact, portable design takes up less floor space
  • DynamicFilter™ self-changing filter system
  • Front fill reservoir
  • Liquid level monitoring to prevent pump failure
  • Ultraviolet anti-biological light system
  • User-adjustable temperature, pressure, and flow rate alarms
Choice of pumps (positive displacement, turbine) and compressor sizes (¼, ⅓, ½, 1 HP).
Certifications: cETLus
Delivery information: Supplied with operating manual, IEC power cord (select models), two sets of inlet/outlet adapters: ½" male NPT×½" hose barb and ½" male NPT×⅝" hose barb (select models).
Caution: Always connect the power cord on this unit to a grounded (3-prong) power outlet. Make certain that the outlet is the same voltage and frequency as your unit. Never operate the unit with a damaged power cord. Always turn the unit OFF and disconnect Mains power before performing any maintenance or service.