Unisette™ Tissue Processing and Biopsy Cassettes, Simport
Supplier: Simport Scientific
M525-2T M525-3T M525-4T M525-5T M525-6T M525-7T M525-8T M525-9T M525-10T M525-11T M525-12T M526-2T M526-3T M526-4T M526-5T M526-6T M526-7T M526-8T M526-9T M526-10T M526-11T M526-12T
76479-292CS 268.77 CAD
76479-292 76479-294 76479-296 76479-298 76479-300 76479-302 76479-304 76479-306 76479-308 76479-310 76479-312 76479-314 76479-316 76479-318 76479-320 76479-322 76479-324 76479-326 76479-328 76479-330 76479-332 76479-334 76479-336 76479-338 76479-340 76479-342 76479-344 76479-346 76479-348 76479-350 76479-352 76479-354 76479-356 76479-358 76479-360 76479-362 76479-364 76479-366 76479-368 76479-370 76479-372 76479-374 76479-376 76479-378
Unisette™ Tissue Processing and Biopsy Cassettes, Simport
Embedding Cassettes Biopsy Processing Cassettes
Our extremely popular Unisette™ histology cassette is now offered with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
  • Made of acetal
  • 45° anterior writing area
  • One-piece integral lid that can be opened and closed up to three times without breakage
  • Secure locking system without danger of specimen loss
  • The biopsy cassette has 1 mm square openings to maximize fluid exchange
  • Offered taped for use with Leica™ IPC printers
Molded from acetal, these cassettes keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap latch and hinge-lock design of the UNISETTE™ prevent early separation of base and lid and allows for one-hand operation.