Eisco LabGlass® Boiling Flasks, Round Bottom, Interchangable Joint
Supplier: Wards
CH0404A CH0404B CH0404C CH0404D CH0404E CH0404F CH0404G CH0404H CH0404I CH0404I01 CH0404J CH0404K CH0404L CH0404M
470331-988EA 9.7 CAD
470331-988 470331-990 470331-992 470331-994 470331-996 470331-998 470332-000 470332-002 470332-004 470332-006 470332-008 470332-010 470332-012 470332-014
Eisco LabGlass® Boiling Flasks, Round Bottom, Interchangable Joint
Flasks Round Bottom Flasks
High quality round bottom boiling flasks with ground, interchangable joint.
  • Made of high quality borosilicate glass 3.3
  • Interchangable, ground joint
  • Stopper not included
  • Round bottom
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.