Anti-DNP HEK293 Polyclonal Antibody

Supplier: ACRO Biosystems
DNPM1A1250TESTS DNPM1A125TESTS
103889-312EA 998.46 CAD
103889-312 103889-314
Anti-DNP HEK293 Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Mouse IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR