Workstation Dispensing Bins
Supplier: Heathrow Scientific
HS23401 HS23402 HS23403 HS23404
76474-350EA 117.27 CAD
76474-350 76474-352 76474-354 76474-356
Workstation Dispensing Bins
Boxes Dispenser Boxes
Clear acrylic bins allow users to see and organize small bulk items such as pipette tips, tubes, fittings, and finger cots.
  • See inventory levels at a glance
  • Flip-top covers make filling and dispensing easy, and keep products dust-free
  • 4 sizes are available to cater to a wide variety of storage needs
  • Includes non-skid rubber feet
The crystal-clear Workstation Dispensing Bin let users see small item inventory with ease. These dispensing bins are made out of sturdy acrylic and come in 4 different sizes. Each bin is proudly made in the USA.
Order Now
SPECIFICATIONS
Environmentally Preferable