Core 6D Carts, TrippNT
Supplier: TrippNT
53463 53464 53465 53468 53474
76471-760EA 2957.26 CAD
76471-760 76471-762 76471-764 76471-766 76471-768 76471-770 76471-772 76471-774 76471-776 76471-778 76471-780 76471-782 76471-784
Core 6D Carts, TrippNT
Trolleys
TrippNT's Core 6D offer 6 total locking drawers for flexible organization and storage.
  • 2 locking and 2 swivel 4" twin wheel casters, 6 full-extension total locking drawers, 6 side shelves (4 shelves, 2 dispensing shelves), tool/chart holder
  • Organizes, stores, and secures supplies
  • 100% total replacement lifetime guarantee/made in USA
It offers a tool holder for tablets, charts, and small utensils. Its built-in handles are ergonomic and easy to grab. It is constructed from a corrosion-resistant non-metal polymer (HDPE) with robust casters capable of easily navigating elevator gaps and uneven surfaces.
Ordering information: Available as fully assembled,13 color variants.