Core DX Carts with Hinged Door, TrippNT
Supplier: TrippNT
53517 53518 53519 53522
76471-840EA 2566.32 CAD
76471-840 76471-842 76471-844 76471-846 76471-848 76471-850 76471-852 76471-854 76471-856 76471-858 76471-860 76471-862 76471-864
Core DX Carts with Hinged Door, TrippNT
Trolleys
TrippNT's Core DX Cart with Hinged Door offers flexible, locking storage and organization for controlled environments.
  • 2 locking and 2 swivel 4" twin wheel casters, 3 total locking drawers, 6 side shelves, locking bulk storage with hinged drawer
  • Built-in easy to grab handles
  • Corrosion-resistant, non-metal HDPE construction
  • Organizes, stores, and secures supplies
  • 100% total replacement lifetime guarantee/made in USA
TrippNT's Core DX Cart organizes, stores, and secures supplies. It offers 3 total locking drawers, 4 shelves, 2 dispensing shelves, andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable