Core DX Carts with Sliding Door, TrippNT
Supplier: TrippNT
53370 53371 53372 53375 53382
76471-708EA 2704.29 CAD
76471-708 76471-710 76471-712 76471-714 76471-716 76471-718 76471-720 76471-722 76471-724 76471-726 76471-728 76471-730 76471-732
Core DX Carts with Sliding Door, TrippNT
Trolleys
TrippNT's Core DX with sliding door offers locking organization and storage for controled environments.
  • 2 locking and 2 swivel 4" twin wheel casters, 3 total locking drawers, 6 side shelves (4 shelves and 2 dispensing shelves), locking bulk storage with a sliding clear door
  • Organizes, stores, and secures supplies
  • 100% total replacement lifetime guarantee/made in USA
Its built-in handles are ergonomic and easy to grab and the sliding door offers a smaller usage footprint. It is constructed from a corrosion-resistant non-metal rust proof polymer (HDPE) with robust casters capable of easily navigating elevator gaps and uneven surfaces.
Ordering information: Available as fully assembled,13 color variants.