Eisco LabGlass® Boiling Flasks, Flat Bottom, Wide Neck
Supplier: Wards
CH0411A CH0411B CH0411C CH0411D CH0411E CH0411F
470332-050EA 6.4 CAD
470332-050 470332-052 470332-054 470332-056 470332-058 470332-060
Eisco LabGlass® Boiling Flasks, Flat Bottom, Wide Neck
Flasks Flat Bottom Flasks
High quality flat bottom boiling flasks with a wide neck design.
  • Made of borosilicate glass 3.3
  • Wide neck design with beaded rim
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.