Sterile Microcentrifuge and Centrifuge Tubes
Supplier: MTC BIO, INC.
C3150-S C2230 C2410 C2600 C2625-R C2625-B C2630 C2602 C2605
470332-378BG 34.75 CAD
470332-378 470332-380 470332-382 470332-384 470332-386 470332-388 470332-390 470332-392 470332-394
Sterile Microcentrifuge and Centrifuge Tubes
Tubes Centrifuge Tubes
These sterile tubes come in a variety of sizes and packaging options to fulfill a variety of tube needs in the lab.
  • RNAse-, DNAse-, pyrogen-, and cytotoxin-free
  • Store at −80 °C
  • Sterilized by gamma radiation
These centrifuge tubes are perfect for everyday sample preparation needs. The rugged construction provides durable, reliable, long-term sample storage.