Eisco LabGlass® Boiling Flasks, Round Bottom with Threaded Joint
Supplier: Wards
CH01002A CH01002B CH01002C CH01002D CH01002E CH01002F CH01002G CH01002H CH01002I CH01002J
470331-772EA 26.95 CAD
470331-772 470331-774 470331-776 470331-778 470331-780 470331-782 470331-784 470331-786 470331-788 470331-790
Eisco LabGlass® Boiling Flasks, Round Bottom with Threaded Joint
Flasks Round Bottom Flasks
High quality round bottom boiling flasks for chemistry classrooms and laboratories.
  • Made of borosilicate glass 3.3
  • Ground threaded joint for secure fit with other glassware
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by aMore Product Information