Eisco LabGlass® Pear Shape Distilling Flasks, Short Neck
Supplier: Wards
CH0407A CH0407B CH0407C CH0407D
470332-024EA 10.1 CAD
470332-024 470332-026 470332-028 470332-030
Eisco LabGlass® Pear Shape Distilling Flasks, Short Neck
Flasks Distillation Flasks
High quality pear shape distilling flasks with ground joints, short neck design.
  • Made of high quality borosilicate glass 3.3
  • Interchangable, ground top joint and side socket
  • Stopper not included
  • Pear shape, short neck design
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of qualityMore Product Information