Corning® HYPERStack® Nest, Corning
Supplier: Corning
Corning®
10047
76478-956CS 404.6 CAD
76478-956
Corning® HYPERStack® Nest, Corning
Racks Bottle Racks
The HYPERStack® Nest accessory is designed for use with the Corning® HYPERStack® 36-layer cell culture vessel.
  • Lightweight design: 2 lbs. (950 g)
  • Simplify process consistency by consistently placing the vessel in the correct position to facilitate optimal fill and empty processes
  • Designed to reduce the chance of pressurizing vessels and wetting filters
  • Pivot angle precisely engineered in the Nest reduces lifting by the user and freeing hands to operate tubing clamps
  • Made of stainless steel, the Nest can be sterilized and autoclaved
  • Small footprint fits within the current HYPERStack® footprint and multiple Nests can be stacked for storage
The HYPERStack® Nest accessory is designed for use with the Corning® HYPERStack® 36-layer cell culture vessel to improve process control by consistently placing the vessel in the optimal positions and angles during use. The HYPERStack® Nest is designed to improve ergonomics by reducing multiple manipulations. Stainless-steel construction allows it to be sterilized and autoclaved, and the small and stackable footprint allows for convenient storage.