Nitrite CRM, Standard Solutions, Supelco®
Supplier: MilliporeSigma
1.25041.0100 1.25042.0100 1.25041.0100 1.25042.0100
EM1.25041.0100CS 4504.58 CAD
EM1.25041.0100 EM1.25042.0100 CA1.25041.0100 CA1.25042.0100
Nitrite CRM, Standard Solutions, Supelco®
Standards Anionic Standards Nitrite Standards
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Certifications: Traceable to SRM from NIST and USP.