STELLARIS 5 Confocal Package, Leica Microsystems

Supplier: Leica Microsystems
Leica®
8119637 8119637
CA76463-390EA 525595.57 CAD
CA76463-390 76463-390
STELLARIS 5 Confocal Package, Leica Microsystems
Microscopes
With STELLARIS, we have re-imagined the confocal microscope, empowering you to drive progress in science.

  • POWER to see more: Power HyD S detectors provide increased sensitivity with true photon counting (power counting) & analog modes
  • PRODUCTIVITY to do more: ImageCompass smart user interface puts you in full control of your experimental setup
  • POTENTIAL to discover more: With just one click LIGHTNING Super Resolution provides simultaneous multicolor super-resolution imaging at the full original speed

STELLARIS 5 Confocal package includes STELLARIS 5 System; 2 Power HyD S detectors; 3 VIS laser Lines (488 nm, 561 nm, 638 nm); 405 nm laser; motorized stage; CS2 PLAN APO objectives (10x and 63x); LAS X STELLARIS Control software (with workstation and monitor); ImageCompass smart user interface; LAS X dye finder; LIGHTNING Super-Resolution; Upgradable to up to 5 detectors; Additional lasers available; Upgradable to DIGITAL LIGHT SHEET (DLS).

Certifications: CE and CAS.

Delivery information: Includes passive microscope table, computer table, and CUDA workstation. Purchase of STELLARIS Confocal Package also includes service installation by a Leica Microsystems field service engineer team.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR