Eisco LabGlass® Distilling Flasks with 3 Parallel Necks, Round Bottom, Interchangable Joints
Supplier: Wards
CH0418A CH0418B CH0418C CH0418D CH0418E CH0418F CH0418G CH0418H CH0418I CH0418J
470332-102EA 48.2 CAD
470332-102 470332-104 470332-106 470332-108 470332-110 470332-112 470332-114 470332-116 470332-118 470332-120
Eisco LabGlass® Distilling Flasks with 3 Parallel Necks, Round Bottom, Interchangable Joints
Flasks Distillation Flasks
High quality round bottom distilling flasks with three parallel necks.
  • Made of borosilicate glass 3.3
  • Ground interchangable joint for secure fit with other glassware
  • Graduated
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.