Purifier® Logic+® and Axiom® Biosafety Cabinet Accessories, Labconco
Supplier: Labconco
3620301 3620302 3620303 7034800 7034801 7034802 7034803 3458401 3458403
76473-118EA 109.43 CAD
76473-118 76473-120 76473-122 76473-124 76473-126 76473-128 76473-130 76473-132 76473-134 76473-136 76473-138 76473-140 76473-142 76473-144 76473-146 76473-148 76473-150 76473-152 76473-154 76473-156
Purifier® Logic+® and Axiom® Biosafety Cabinet Accessories, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Accessories for use with Labconco Class II biosafety cabinets.
  • Accessories for various needs
  • Trace odor filters for nuisance odors
  • Prop rod holds work surface in raised position
BSC sash stop restricts the sash from closing to maintain continuous operation. For use with Labconco Class II biosafety cabinets. The dry contact relay kit for use with L3 generation Logic+ and Axiom biosafety cabinets.
Easy-clean prop rod holds the work surface in a raised position for easy cleaning of the sump area on Logic+ BSCs. Short version for use with service-fixtured BSCs. Long version for use with non-fixtured BSCs.
Replacement trace odor filter for Logic+ Type A2 trace odor filter kits. For use with Logic+ Type A2 biosafety cabinets. Trace odor filter kit for Logic+ A2 biosafety cabinets. Removes trace nuisance odors from the exhaust air of Logic+ Class II, Type A2 BSCs. Contact us for applications involving light-duty chemicals.
BSC flex duct for connection of ductwork to BSC exhaust damper. 10" Nominal diameter. Includes flex duct length and 2× stainless steel band clamps.
Axiom flat work surface. Flat replacement for standard dished Chem-Zone work surface. For use with Axiom Class II Type C1 biosafety cabinets.