Leaf and Flower Press
Supplier: Ward's Science
506.4001.1 506.4021.50 506.4023.50 506.4031.2
470332-434EA 65.4 CAD
470332-434 470332-440 470332-454 470332-462
Leaf and Flower Press
Presses
Extra strength for thicker samples.
  • Small size for field collection
  • Strapping provides heavier compression
Ideal for field work because of its portable size, 6×9". This plant press is frequently used by Ecologists to gather base-line data for impact studies. Also used by Botanists and Entomologists in host plant studies. Ruggedly constructed of ¼" plywood strengthened by hardwood side strips which provide uniform pressure.
Delivery information: Plant press comes complete with 12 drying sheets, 12 corrugated ventilators, and two straps with buckles.
Order Now
SPECIFICATIONS
Environmentally Preferable