Eisco LabGlass® Erlenmeyer Flasks with Polypropylene Screw Cap
Supplier: Wards
CH0430A CH0430B CH0430C CH0430D CH0430E CH0430F
470332-208EA 12.3 CAD
470332-208 470332-210 470332-212 470332-214 470332-216 470332-218
Eisco LabGlass® Erlenmeyer Flasks with Polypropylene Screw Cap
Flasks Erlenmeyer Flasks
High quality Erlenmeyer flask with polypropylene screw cap for chemistry classrooms and laboratories.
  • Made of high quality borosilicate glass 3.3
  • Includes acid proof polypropylene screw cap
  • Graduated
  • Fully autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each productMore Product Information