Eisco LabGlass® Reagent Bottles with Hexagonal Glass Stopper, Wide Mouth
Supplier: Wards
CH0163A CH0163B CH0163C CH0163D CH0163E CH0163F
470331-934EA 15 CAD
470331-934 470331-936 470331-938 470331-940 470331-942 470331-944
Eisco LabGlass® Reagent Bottles with Hexagonal Glass Stopper, Wide Mouth
Bottles Glass Bottles
High-quality glass reagent bottle useful for storing chemicals in classrooms or laboratories.
  • Made of high-quality borosilicate glass 3.3
  • Interchangeable, ground glass stopper
  • Ungraduated
  • Wide mouth design
  • Fully autoclavable
  • Superior chemical durability
  • Amber colored glassware protects light-sesitive chemicals
Low temperature gradient - can withstand high temperatures and thermal shocks. Heat tolerances: Strain point 515 °C; annealing point 565 °C; softening point 820 °C.