Eisco LabGlass® Distilling Flasks with Side Arm, Round Bottom
Supplier: Wards
CH0416A CH0416B CH0416C CH0416D CH0416E
470332-092EA 17.4 CAD
470332-092 470332-094 470332-096 470332-098 470332-100
Eisco LabGlass® Distilling Flasks with Side Arm, Round Bottom
Flasks Distillation Flasks
High quality round bottom distilling flasks with side arm.
  • Made of high quality borosilicate glass 3.3
  • Beaded rim with side arm
  • Round bottom
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals andMore Product Information