Eisco LabGlass® Pear Shape Boiling Flasks, Interchangable Joint
Supplier: Wards
CH0406A CH0406B CH0406C CH0406D
470332-016EA 8.85 CAD
470332-016 470332-018 470332-020 470332-022
Eisco LabGlass® Pear Shape Boiling Flasks, Interchangable Joint
Flasks Round Bottom Flasks
High quality pear shapeboiling flasks with ground, interchangable joint.
  • Made of high quality borosilicate glass 3.3
  • Interchangable, ground joint
  • Stopper not included
  • Pear shape design
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.