Eisco LabGlass® Distillation Flasks, 3 Necks with Threaded Joint
Supplier: Wards
CH01010A CH01010B CH01010C CH01010D CH01010E CH01010F
470331-844EA 86.6 CAD
470331-844 470331-846 470331-848 470331-850 470331-852 470331-854
Eisco LabGlass® Distillation Flasks, 3 Necks with Threaded Joint
Flasks Distillation Flasks
High quality distillation flask for chemistry classrooms and laboratories.
  • Made of high quality borosilicate glass 3.3
  • Ground threaded joints for secure fit with other glassware
  • 3 neck design (angled)
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product isMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable