Eisco LabGlass® Boiling Flasks, Flat Bottom with Threaded Joint
Supplier: Wards
CH01004A CH01004B CH01004C CH01004D CH01004E CH01004F CH01004G CH01004H CH01004I CH01004J
470331-792EA 24.15 CAD
470331-792 470331-794 470331-796 470331-798 470331-800 470331-802 470331-804 470331-806 470331-808 470331-810
Eisco LabGlass® Boiling Flasks, Flat Bottom with Threaded Joint
Flasks Flat Bottom Flasks
High quality flat bottom boiling flasks for chemistry classrooms and laboratories.
  • Made of high quality borosilicate glass 3.3
  • Ground threaded joint for secure fit with other glassware
  • Flat bottom design
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each productMore Product Information