Eisco LabGlass® Boiling Flasks, Flat Bottom, Interchangeable Joint
Supplier: Wards
CH0412A CH0412B CH0412B01 CH0412B02 CH0412C CH0412D CH0412E CH0412F CH0412G CH0412H CH0412I CH0412I01 CH0412J CH0412K CH0412L
470332-062EA 8.85 CAD
470332-062 470332-064 470332-066 470332-068 470332-070 470332-072 470332-074 470332-076 470332-078 470332-080 470332-082 470332-084 470332-086 470332-088 470332-090
Eisco LabGlass® Boiling Flasks, Flat Bottom, Interchangeable Joint
Flasks Flat Bottom Flasks
High quality flat bottom boiling flasks with ground, interchangeable joint.
  • Made of high quality borosilicate glass 3.3
  • Interchangeable, ground joint
  • Stopper not included
  • Flat bottom
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment and following the highest standards of quality control. Each product is carefully designedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable