Eisco LabGlass® Graham Condensers, Coiled Distillate
Supplier: Wards
CH0308HN CH0308JN
470331-952EA 67.8 CAD
470331-952 470331-954
Eisco LabGlass® Graham Condensers, Coiled Distillate
Condensers
High quality glass Graham condenser with coiled distillate.
  • Made of high quality borosilicate glass 3.3
  • Ground socket and cone for a secure connection with other glassware
  • Two tapered side arms for connecting tubing securely
  • Coiled distillate design
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.