EiscoLabGlass® Vigreux Columns with Threaded Joint
Supplier: Wards
CH01016A CH01016B CH01016C
470331-872EA 93.55 CAD
470331-872 470331-874 470331-876
EiscoLabGlass® Vigreux Columns with Threaded Joint
Distillation Components Distillation Columns
High quality Vigreux column used to separate vaporized mixtures of liquid compounds with close volatility.
  • Made of high quality borosilicate glass 3.3
  • Ground threaded joints for secure fit with other glassware
  • Ground cone for secure fit with other glassware
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.