Eisco LabGlass® Liebig Condensers with Threaded Joint
Supplier: Wards
CH01012A CH01012B CH01012C CH01012D
470331-856EA 68.4 CAD
470331-856 470331-858 470331-860 470331-862
Eisco LabGlass® Liebig Condensers with Threaded Joint
Condensers
High quality Liebig condenser for chemistry classrooms and laboratories.
  • Made of high quality borosilicate glass 3.3
  • Ground threaded joints for secure fit with other glassware
  • Ground cone for secure fit with other glassware
  • Two tapered side arms for connecting tubing securely
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.