HyClone™ ActiPro Cell Culture Medium, HyClone products (Cytiva)
Supplier: Cytiva
CA76462-566EA 464.46 CAD
CA76462-566 CA76462-568
HyClone™ ActiPro Cell Culture Medium, HyClone products (Cytiva)
Cell Culture Media
HyClone™ ActiPro cell culture medium has been formulated to provide high yields of recombinant proteins in processes using Chinese hamster ovary (CHO) cell lines.
  • Demonstrated high yields of recombinant proteins
  • Does not contain hypoxanthine and thymidine
  • Chemically defined ADCF formulation
  • Production medium for batch or fed-batch processes
  • Accompanied with detailed protocols for use to maximize output
HyClone™ ActiPro does not contain any growth factors such as insulin, peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batch consistency and increased cell culture process efficiency.