HyClone™ ActiSM Cell Culture Medium, HyClone products (Cytiva)
Supplier: Cytiva
SH31038.01
CA76462-570EA 604.89 CAD
CA76462-570 CA76462-572
HyClone™ ActiSM Cell Culture Medium, HyClone products (Cytiva)
Cell Culture Media
HyClone™ ActiSM cell culture media is a chemically defined animal-derived component-free (ADCF) cultured media optimized for high-yield protein production for bioprocessing. This media is designed for use with HyClone™ ActiPro cell culture media.
  • Does not contain hypoxanthine, thymidine, and glycine (HAT supplement)
  • Recommended for first step when adapting cells cultured in other lean cell culture media
  • Storage temperature 2 to 8 °C and storage condition is away from light and moisture
  • Additive: with Poloxamer 188; without L-glutamine
HyClone™ ActiSM medium does not contain any growth factors such as insulin, peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring CHO batch-to-batch consistency.
Cell culture media formulations and delivery systems can be customized to specific process requirements or optimized to maximize process yields. Our cell culture media and development service is available to help to analyze, develop, and optimise cell culture strategies.