Arium® Smart Station Pure Water Remote Dispenser, Sartorius
Supplier: Sartorius
H2O-ARST-P-T H2O-ARST-P-B H2O-ARST-P-T H2O-ARST-P-B
CA76474-080EA 2701 CAD
CA76474-080 CA76474-082 76474-080 76474-082
Arium® Smart Station Pure Water Remote Dispenser, Sartorius
Water Dispensers
The Arium® Smart Station can be optimized to fit easily into any lab environment. Combine the Smart Station with the water system type needed to dispense pure water. The Smart Station pure supports multiple points of use to save costs and valuable lab space.
  • Compact: Save space integrating into any lab
  • Intuitive: Touch-activated color display with direct access to all important functions
  • Flexible: Stepless height adjustment to fill different size containers
  • Ergonomic design: Hand-held for optimized left or right-hand operation
  • Accurate: Precise volume
More Product Information