Arium® Smart Station Ultrapure Water Remote Dispenser, Sartorius
Supplier: Sartorius
H2O-ARST-UP-B H2O-ARST-UP-B H2O-ARST-UP-T
CA76474-078EA 3219.34 CAD
CA76474-078 76474-078 CA76474-842
Arium® Smart Station Ultrapure Water Remote Dispenser, Sartorius
Water Dispensers
The Arium® Smart Station can be optimized to fit easily into any lab environment. Combine the Smart Station with the water system type needed to dispense pure or ultrapure water. The Smart Station ultrapure supports multiple points of use to save costs and valuable lab space.
  • Compact: Save space integrating into any lab
  • Intuitive: Touch-activated color display with direct access to all important functions
  • Flexible: Stepless height adjustment to fill different size containers
  • Ergonomic design: Hand-held for optimized left or right-hand operation
  • Accurate: Precise volume dispense for reliable buffer and sample preparation
Flexible remote dispenser for Arium® Pro and Comfort systems. Height adjustable display/dispense station for dispensing Type 1 ultrapure water.
Intended for use with Arium®Pro DI, Pro UF, Pro UV, Pro VF, and Arium® Comfort.
Certifications: CE declaration of conformity.
Delivery information: Equipment supplied: Display/dispense unit with stand, power supply including IEC connector, connection set.