Motic Moticam S Series, Microscope Cameras
Supplier: National Optical
D-MOTICAM S1 D-MOTICAM S3 D-MOTICAM S6 D-MOTICAMS12
470332-264EA 733.1 CAD
470332-264 470332-266 470332-268 470332-270
Motic Moticam S Series, Microscope Cameras
Cameras
Ideal for in-person or distance learning and instruction.
  • Fast frame rates
  • sCMOS sensor
  • USB 3.1 interface
Motic's new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. Easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
Ordering information: Included Motic Images software enables live and captured images atMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable