HyClone Cell Boost™ 7b Supplement, HyClone products (Cytiva)
Supplier: Cytiva
SH31120.01 SH31120.02 SH31120.03 SH31027.07 SH31027.01
CA76462-602EA 107.01 CAD
CA76462-602 CA76462-604 CA76462-606 CA76462-556 CA76534-644
HyClone Cell Boost™ 7b Supplement, HyClone products (Cytiva)
Cell Culture Supplements and Additives
Supplements cell culture with concentrated amino acids. Manufactured to meet cGMP manufacturing standards and QC specifications.
  • Designed to support modern manufacturing processes, Cell Boost 7b supplement is a concentrated solution of amino acids with an alkaline pHSupplement is chemically defined, animal-derived component-free (ADCF), do not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Use Cell Boost 7a and 7b together to achieve high yields of mAbs and other proteins in CHO fed-batch processes
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
  • Non sterile
Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines. Cell Boost 7b is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.