Stuart® Magnetic Hotplate Stirrers, Digital, Undergrad Series, Cole-Parmer
Supplier: Cole Parmer
stuart®
04807-65 04807-63
76474-332EA 2064.37 CAD
76474-332 76474-330
Stuart® Magnetic Hotplate Stirrers, Digital, Undergrad Series, Cole-Parmer
Stirrers Magnetic Hotplate Stirrers
Get true temperature setting and accurate stirring control.
  • Hot warning light remains on even when unplugged
  • BioCote® glaze for antimicrobial protection
  • Tough aluminum casing for longevity
  • Designed to prevent accidental spillage on control panel
  • Compact, stackable design takes up less benchtop and storage space
  • IP 32 rated for water and dust resistance
Designed with microprocessor-controlled dual thermocouples, these magnetic hotplate stirrers offer accurate temperature control and protection from over-heating. Actual surface temperature is displayed in real time on an easy-to-read digital screen. Powerful magnets and motor provide stirring speeds of up to 2000 rpm and mixing capability of up to 15 L of liquid. The square plate design allows flexibility of vessel sizes and combinations. Up to four 250 ml beakers can easily fit on the surface at the same time.
To keep the user safe, a “hot” warning light flashes whenever the plate surface temperature is above 50 °C and continues to flash until the temperature drops below 50 °C, even when unplugged. The units also include BioCote® antimicrobial protection which supports a safer and more hygienic laboratory environment, while reducing risk of cross contamination in the laboratory.
All models include an integral fitting for a retort rod and allow the retort base to slide underneath. For convenience and power saving, the units are built with a standard mains switch.
Available with either coated aluminum top plate for rapid heating and better temperature uniformity or chemically resistant glass ceramic that stays cool around the edges to help prevent burns. Compatible with the Stuart® SCT2 temperature controller.
Certifications: IP protection class according to DIN EN 60529: IP 32