Defender™ 5000 Industrial Bench Scale
Supplier: Ohaus
30461655 30461642 30461660 30461645 30467607 30461639 30461657 30461630 30461648 30461633 30461651 30461643 30467606 30461646 30461640 30461658 30461631 30461649 30461634 30461652 30461644 30461647 30461636 30461654 30461632 30461650 30461641 30461659 30461635 30461653 30461638 30461656 30461637
76472-636EA 3770.34 CAD
76472-636 76472-638 76472-640 76472-642 76472-644 76472-646 76472-648 76472-650 76472-652 76472-654 76472-656 76472-658 76472-660 76472-662 76472-664 76472-666 76472-668 76472-670 76472-672 76472-674 76472-676 76472-678 76472-680 76472-682 76472-684 76472-686 76472-688 76472-690 76472-692 76472-694 76472-696 76472-698 77434-854
Defender™ 5000 Industrial Bench Scale
Balances and Scales
The OHAUS next generation Defender 5000 Series multifunctional bench scales are ideal for a multitude of applications, including production, packaging, inventory and shipping. Durable and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options - the Defender 5000 series is designed to simplify demanding industrial and commercial applications.
  • Indicator guides user with on-screen prompts
  • Durable construction to withstand harsh industrial environments
  • Multiple application modes and high storage capacity for increased flexibility
  • Multiple connectivity options for convenient data transfer and storage
  • Backlit LCD display, 256×80 pixels
  • Durably built
  • Defender 5000 Series is designed to simplify demanding industrial and commercial applications
  • Equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options
Communication is facilitated by means of the easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, discrete I/O and Analog Output (sold separately as accessories) and GLP/GMP data output with real-time clock.
The TD52P indicator is made from ABS plastic and the TD52XW indicator is stainless steel, scales have a 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell and non-slip adjustable rubber feet.
Applications include: weighing, parts counting, percent weighing, check weighing, dynamic weighing (display hold). Units: custom; gram; kilogram; metric tonnes; ounce; pound; pound: ounce and tonne.
Design features include user-configurable resolution up to dual range 15000d/3000e (25000d/5000d for NTEP), 150% overload capacity protection, check weighing symbols with selectable operation and audible signal settings, menu lock switch, multiple selectable operating languages, selectable environmental and auto-print settings, stability indicator, overload/underload indicators, selectable adjust contrast settings, auto-dim display, auto-off, brightness adjustment, screensaver and auto tare.
Certifications: Metrology: Measurement Canada (Indicator: NoA AM-6093, Platforms: NoA AM-6107); NTEP (Indicator: CC 18-065, Platforms: CC 18-077) Class lll
Product Safety: CAN/CSA C22.2 61010-1; UL61010-1; IEC/EN 61010-1
Electromagnetic Compatibility: FCC Part 15 Class B; Canada ICES-003 Class B; IEC/EN 61326-1 Class B, basic environments, Compliance Marks: CE; CSA
The defender complies with NTEP/Measurement Canada or OIML/EC approvals (depending on the region)
Ordering information: Scales have an internal power supply and are supplied with a power cord or can be used with an optional rechargeable battery (sold separately as an accessory).