Benchtop Centrifuges and Packages, Multifuge X1 and X1R Pro, Thermo Scientific
Supplier: Thermo Fisher Scientific
75009210 75009251 75016008 75017008 75016024US 75017024US 75009210CAP 75009251CAP 75016008CAP 75017008CAP 75016024USCAP 75017024USCAP
76470-412EA 17103.28 CAD
76470-412 76470-416 76470-440 76470-444 76470-452 76470-456 CA76501-570 CA76501-572 CA76501-608 CA76501-616 CA76501-606 CA76501-614
Benchtop Centrifuges and Packages, Multifuge X1 and X1R Pro, Thermo Scientific
Centrifuges Benchtop Centrifuges
Ergonomically designed, the Thermo Scientific™ Multifuge X1/X1R Pro series benchtop centrifuges have a touch screen interface that enables easier programming for quicker results and ability to access and track all runs, rotor life and centrifuge health information for maximum versatility and power at every turn.
  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life. Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.
Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. Technology optimizes safety and improves separations.
Compact benchtop dimensions optimize working area, the ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Certifications: IVD certified.
Ordering information: Centrifuge only, optional Cell Culture and Blood Tube packages are also available.
Cell Culture package includes TX400 rotor, set of four 400 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 4×50 ml conical tube adapters, set of four 9×15 ml conical tube adapters.
Blood Tube package includes TX400 rotor, set of four 400 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 19×5/7 ml blood tube and 14×10 ml blood tube adapters.