Thermo Scientific™ Floor Standing Centrifuges and Packages, Multifuge X4F and X4FR Pro, Thermo Scientific
Supplier: Thermo Scientific
75009537 75016048 75016047 75009531
76470-408EA 28559.61 CAD
76470-408 76470-434 76470-410 76470-432
Thermo Scientific™ Floor Standing Centrifuges and Packages, Multifuge X4F and X4FR Pro, Thermo Scientific
Centrifuges Floor-Standing Centrifuges
Ergonomically designed, the Thermo Scientific™ Multifuge X4F/X4FR Pro series floor-standing centrifuges have a touch screen interface that enables easier programming for quicker results and ability to access and track all runs, rotor life and centrifuge health information for maximum versatility and power at every turn.
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.
Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. Technology optimizes safety and improves separations.
Floor standing option and compact dimensions optimize working area, the ergonomic height and curved design with motorized lid latch ensure easy sample loading and unloading, easy cleaning and promotes user safety.
System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Certifications: IVD certified.
Ordering information: Centrifuge only, optional Cell Culture and Blood Tube packages are also available.
Cell Culture package includes TX1000 rotor, set of four 1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 10×50 ml conical tube adapters, set of four 24×15 ml conical tube adapters.
Blood Tube package includes TX1000 rotor, set of four 1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 49×5/7 ml blood tube and 37×10 ml blood tube adapters.