Thermo Scientific™ Floor Standing Centrifuges and Packages, Megafuge ST4F and ST4FR Plus, Thermo Scientific
Supplier: Thermo Fisher Scientific
75009946 75009960 75016059 75017946 75017960 75017964 75009960CAP 75009946CAP 75017960CAP 75017946CAP 75016059CAP
76470-404EA 29391.7 CAD
76470-404 76470-406 76470-436 76470-446 76470-448 76470-450 CA76501-582 CA76501-584 CA76501-622 CA76501-624 CA76501-630 CA76501-632
Thermo Scientific™ Floor Standing Centrifuges and Packages, Megafuge ST4F and ST4FR Plus, Thermo Scientific
The Thermo Scientific™ Megafuge ST4F/ST4FR Plus floor-standing centrifuges offer intuitive controls and versatility for power at every turn.
  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.
Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. Technology optimizes safety and improves separations.
Floor standing option and compact dimensions optimize working area, the ergonomic height and curved design with motorized lid latch ensure easy sample loading and unloading, easy cleaning and promotes user safety.
Certifications: IVD certified.
Ordering information: Centrifuge only, optional Cell Culture and Blood Tube packages are also available.
Cell Culture package includes TX1000 rotor, set of four 1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 10×50 ml conical tube adapters, set of four 24×15 ml conical tube adapters.
Blood Tube package includes TX1000 rotor, set of four 1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 49×5/7 ml blood tube and 37×10 ml blood tube adapters.