Vials with Anticoagulant Additive for Blood Sample Collection, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
HL100 ECV-2.5 ECVK2-0.8-25
76452-020PK 721.84 CAD
76452-020 76452-022 76452-024
Vials with Anticoagulant Additive for Blood Sample Collection, SAI Infusion Technologies
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These borosilicate glass vials are a perfect fit for certain automated blood sampling systems. Available with pharmaceutical grade heparin, EDTA, citrate, or your choice of anticoagulant. Formulated for various blood volumes with a variety of additives. Can be customized.
  • Maximize plasma or serum recovery from the smallest blood samples
  • Anticoagulant type and amount are customizable
  • Provided sterile
These extremely small (6×32 mm) borosilicate glass vials provide maximum plasma or serum recovery from the smallest blood samples, from 10 to 300 µl.More Product Information