CyBio® FeliX Automated Pipet Robots, Analytik Jena US

Supplier: Analytik Jena CA
OL5015-24-100 OL5015-25-500 OL5015-25-505
CA76469-566EA 60487.23 CAD
CA76469-566 CA76469-568 CA76469-570
CyBio® FeliX Automated Pipet Robots, Analytik Jena US
Automated Liquid Handling Systems
Flexible pipetting platform for fully automated single to multichannel liquid handling.

  • Flexible pipettor with 1 to 384 channels with automatic loading of pipetting tools
  • Unique and compact deck design, 12 positions on two levels
  • CHOICE™ technology for pipetting from 500 nl to 1 ml
  • Highest precision and accuracy for various applications in all formats
  • Closed stand-alone system or as an open system for placing in laminar flow hood and integration

The CyBio® FeliX offers maximum performance for the automation of your cellular assays such as media exchange, cell seeding, ADME/Tox, transfection and cell-based screening. The precision and reliable functionality of the CyBio® FeliX was exemplified in the automation of a Cisbio HTRF® cellular phospho-assay.

CyBio® FeliX is a flexible and compact liquid handling platform for single to multichannel pipetting on the smallest footprint of 650×450 mm. HTRF® cellular kinase phospho-assays provide an ideal phosphodetection readout enabling screening of signaling processes in cells – easily automatable on the CyBio® FeliX.

With configurable accessories and interchangeable pipetting heads and tools for 1- to 384-channel pipetting, CyBio® FeliX can process even more laborious tasks like plate replication, creating reaction set-ups for PCR and Next Gen Sequencing, ELISA and serial dilution.

CyBio®-FeliX – automated application starts here:
- Feel from work-intense, manual pipetting - with fully automated pipetting in different formats and integrated tool and tip exchange
- Increase your reproducibility and save reagent costs - with CyBio's proven pipetting technology and liquid specific parameters for highest precision and accuracy
- Save valuable lab space - maximum performance with minimal dimensions of 650×450×700 mm (W×D×H)
- Be prepared for the future - with the reliable and easy to adapt CyBio®-FeliX system

Discover CHOICE™ technology
The CHOICE™ technology overcomes the limits of today's pipetting technology. Using only one pipetting head, the patent pending design allows for:
- Extensive volume range: the head design results an unique overall volume range of 500 nl to 1 ml combined in one pipetting head
- Enhanced format flexibility: supports 9 mm pitch (96 well format) and 4.5 mm pitch (384 well format) in single, row and column mode by loading of appropriate CHOICE™ adapters and tips without manual intervention
- After pickup, each adapter is automatically identified, with optimized pipetting parameters for highly precise and reproducible liquid handling results

Ordering information: Pipetting heads and consumables for the CyBio® FeliX are sold separately.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR