Vaccination Labels 'Contains Vaccine, Do Not Unplug', Accuform
Supplier: ACCUFORM MANUFACTURING, INC
MBDX508MG MBDX505VS MBDX505XV MBDX505MG
76471-308EA 10.81 CAD
76471-308 76471-310 76471-312 76471-314 76471-316 76471-318
Vaccination Labels 'Contains Vaccine, Do Not Unplug', Accuform
Labels
Vaccination Labels communicate health and safety policies, reinforce procedures, and identify locations.
  • Multiple material, size, and color options are available
Choose from three material options. Adhesive Vinyl lables can be applied to curved surfaces such as pipes, poles, and secondary containers. They are moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. The printed matte surface has a permanent acrylic adhesive back, and the finishing includes square corners and scored back liner for quick peel-off and application.
Adhesive Dura-Vinyl lables feature a printed image protected by a durable UV coating for indoor and outdoor marking locations. Permanent acrylic adhesive back. Dual-layer flexibility can be applied to curved areas such as pipes, poles, and secondary containers. Round corners and scored back liner for quick peel-off and application.
Magnetic Vinyl lables feature flexible vinyl that mounts to most steel or ferrous metal surfaces. Easily lift, move, and replace. Message is surface printed and laminated to resist chipping and cracking.