Signs, 'EXIT', Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
MADC531VS MADC531VP MADC531VA MEXT906VS MEXT906VP MEXT906VA
76471-296EA 15.18 CAD
76471-296 76471-298 76471-300 76471-302 76471-304 76471-306
Signs, 'EXIT', Accuform®
Signs
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Multiple material and size options available
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. These are moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Plastic is a good material choice for indoor and sheltered outdoor environments, as the printed surface has clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. They feature a printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions. They also offer abrasion and chemical resistance.